| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100005285.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005285.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005285.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005284.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005284.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005284.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005287.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005287.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005287.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005280.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005280.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005280.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005289.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005289.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005289.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005281.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005281.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005281.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005288.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005288.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005288.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005282.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005282.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005282.mm.1 |
TC0100000640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100005286.mm.1 |
NM_001014974.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005286.mm.1 |
NM_001014974.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100005286.mm.1 |
TC0100000640.mm.1 |
MTA TC |