Assay Details
| Gene Symbol: Snapc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029949.3 | NP_084225.1 | 4 - 5 | 583 | 116 | |
| GenBank mRNA | AK007563.1 | - | 4 - 5 | 583 | 116 | |
| AK010932.1 | - | 4 - 5 | 584 | 116 | ||
| AK016168.1 | - | 4 - 5 | 598 | 116 | ||
| AK019863.2 | - | 4 - 5 | 579 | 116 | ||
| AK031870.1 | - | 4 - 5 | 585 | 116 | ||
| AK049933.1 | - | 4 - 5 | 604 | 116 | ||
| BC048713.1 | - | 4 - 5 | 583 | 6330194 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snapc3
Genomic Map