Assay Details
| Gene Symbol: Snapc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029949.3 | NP_084225.1 | 6 - 7 | 807 | 125 | |
| GenBank mRNA | AK007563.1 | - | 6 - 7 | 807 | 125 | |
| AK019863.2 | - | 6 - 7 | 803 | 125 | ||
| AK031870.1 | - | 6 - 7 | 809 | 125 | ||
| AK031924.1 | - | 5 - 6 | 725 | 125 | ||
| AK049933.1 | - | 6 - 7 | 828 | 125 | ||
| BC048713.1 | - | 6 - 7 | 807 | 6330194 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snapc3
Genomic Map