Assay Details
| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045528.1 | NP_001038993.1 | 25 - 26 | 2886 | 129 | |
| GenBank mRNA | AK122343.1 | - | 18 - 19 | 2125 | 129 | |
| AK170070.1 | - | 5 - 6 | 631 | 129 | ||
| BC034098.1 | - | 15 - 16 | 1823 | 129 | ||
| BC060651.1 | - | 25 - 26 | 2800 | 129 | ||
| BC137706.1 | - | 25 - 26 | 2790 | 8860725 | 129 | |
More Information
Set Membership: |
|

Fkbp15
Genomic Map