Assay Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009554.4 | NP_033580.3 | 1 - 2 | 215 | 77 | |
| GenBank mRNA | AK032469.1 | - | 1 - 2 | 97 | 77 | |
| BC063757.1 | - | 1 - 2 | 99 | 6331110 | 77 | |
| CO430987.1 | - | 1 - 2 | 97 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032469.1, BC063757.1, NM_009554.4 transcripts.
Set Membership: |
|

Zfp37
Genomic Map