Assay Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290521.1 | NP_001277450.1 | 2 - 3 | 279 | 77 | |
| NM_001290522.1 | NP_001277451.1 | 2 - 3 | 172 | 77 | ||
| NM_023597.2 | NP_076086.1 | 2 - 3 | 172 | 77 | ||
| GenBank mRNA | AF274321.1 | - | 2 - 3 | 149 | 77 | |
| AK017259.1 | - | 2 - 3 | 165 | 77 | ||
| AK141026.1 | - | 2 - 3 | 277 | 77 | ||
| BC026500.1 | - | 1 - 2 | 58 | 77 | ||
| BC037102.1 | - | 2 - 3 | 162 | 5364307 | 77 | |
| CK618879.1 | - | 2 - 3 | 279 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF274321.1, AK017259.1, AK141026.1, BC037102.1, NM_001290521.1, NM_001290522.1, NM_023597.2 transcripts.
Set Membership: |
|

Wdr31
Genomic Map