Assay Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290521.1 | NP_001277450.1 | 3 - 4 | 426 | 76 | |
| NM_023597.2 | NP_076086.1 | 3 - 4 | 319 | 76 | ||
| GenBank mRNA | AF274321.1 | - | 3 - 4 | 296 | 76 | |
| AK141026.1 | - | 3 - 4 | 424 | 76 | ||
| BC026500.1 | - | 2 - 3 | 205 | 76 | ||
| BC037102.1 | - | 3 - 4 | 309 | 5364307 | 76 | |
| CK618879.1 | - | 3 - 4 | 426 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr31
Genomic Map