Assay Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290521.1 | NP_001277450.1 | 5 - 6 | 631 | 60 | |
| NM_001290522.1 | NP_001277451.1 | 5 - 6 | 521 | 60 | ||
| NM_023597.2 | NP_076086.1 | 5 - 6 | 524 | 60 | ||
| GenBank mRNA | AF274321.1 | - | 5 - 6 | 501 | 60 | |
| AK017259.1 | - | 5 - 6 | 514 | 60 | ||
| AK141026.1 | - | 5 - 6 | 629 | 60 | ||
| BC026500.1 | - | 4 - 5 | 410 | 60 | ||
| BC037102.1 | - | 5 - 6 | 514 | 5364307 | 60 | |
More Information
Set Membership: |
|

Wdr31
Genomic Map