Assay Details
| Gene Symbol: Prpf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027297.3 | NP_081573.1 | 11 - 12 | 1237 | 73 | |
| GenBank mRNA | AK005464.1 | - | 11 - 12 | 1216 | 73 | |
| AK085174.1 | - | 11 - 12 | 1201 | 73 | ||
| AK089838.1 | - | 11 - 12 | 1189 | 73 | ||
| AK090114.1 | - | 11 - 12 | 1214 | 73 | ||
| AK166912.1 | - | 11 - 12 | 1223 | 73 | ||
| AK167216.1 | - | 11 - 12 | 1215 | 73 | ||
| AK169532.1 | - | 11 - 12 | 1200 | 73 | ||
| BC051639.1 | - | 7 - 8 | 651 | 73 | ||
| BC106089.1 | - | 11 - 12 | 1199 | 30715403 | 73 | |
| BC125529.1 | - | 11 - 12 | 1188 | 40130044 | 73 | |
| BC125531.1 | - | 11 - 12 | 1188 | 40130046 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf4
Genomic Map