Assay Details
| Gene Symbol: Prpf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027297.3 | NP_081573.1 | 2 - 3 | 301 | 72 | |
| GenBank mRNA | AK005464.1 | - | 2 - 3 | 280 | 72 | |
| AK085174.1 | - | 2 - 3 | 264 | 72 | ||
| AK089838.1 | - | 2 - 3 | 253 | 72 | ||
| AK090114.1 | - | 2 - 3 | 278 | 72 | ||
| AK166912.1 | - | 2 - 3 | 287 | 72 | ||
| AK167216.1 | - | 2 - 3 | 279 | 72 | ||
| AK169532.1 | - | 2 - 3 | 264 | 72 | ||
| BC106089.1 | - | 2 - 3 | 263 | 30715403 | 72 | |
| BC125529.1 | - | 2 - 3 | 252 | 40130044 | 72 | |
| BC125531.1 | - | 2 - 3 | 252 | 40130046 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf4
Genomic Map