Assay Details
| Gene Symbol: Prpf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027297.3 | NP_081573.1 | 3 - 4 | 480 | 81 | |
| GenBank mRNA | AK005464.1 | - | 3 - 4 | 459 | 81 | |
| AK031219.1 | - | 466 | 81 | |||
| AK085174.1 | - | 3 - 4 | 443 | 81 | ||
| AK089838.1 | - | 3 - 4 | 432 | 81 | ||
| AK090114.1 | - | 3 - 4 | 457 | 81 | ||
| AK166912.1 | - | 3 - 4 | 466 | 81 | ||
| AK167216.1 | - | 3 - 4 | 458 | 81 | ||
| AK169532.1 | - | 3 - 4 | 443 | 81 | ||
| BC106089.1 | - | 3 - 4 | 442 | 30715403 | 81 | |
| BC125529.1 | - | 3 - 4 | 431 | 40130044 | 81 | |
| BC125531.1 | - | 3 - 4 | 431 | 40130046 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf4
Genomic Map