Assay Details
| Gene Symbol: Prpf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027297.3 | NP_081573.1 | 6 - 7 | 740 | 81 | |
| GenBank mRNA | AK005464.1 | - | 6 - 7 | 719 | 81 | |
| AK031219.1 | - | 726 | 81 | |||
| AK089838.1 | - | 6 - 7 | 692 | 81 | ||
| AK090114.1 | - | 6 - 7 | 717 | 81 | ||
| AK166912.1 | - | 6 - 7 | 726 | 81 | ||
| AK167216.1 | - | 6 - 7 | 718 | 81 | ||
| AK169532.1 | - | 6 - 7 | 703 | 81 | ||
| BC051639.1 | - | 2 - 3 | 154 | 81 | ||
| BC106089.1 | - | 6 - 7 | 702 | 30715403 | 81 | |
| BC125529.1 | - | 6 - 7 | 691 | 40130044 | 81 | |
| BC125531.1 | - | 6 - 7 | 691 | 40130046 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf4
Genomic Map