Assay Details
| Gene Symbol: Prpf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027297.3 | NP_081573.1 | 9 - 10 | 1027 | 71 | |
| GenBank mRNA | AK005464.1 | - | 9 - 10 | 1006 | 71 | |
| AK085174.1 | - | 9 - 10 | 991 | 71 | ||
| AK089838.1 | - | 9 - 10 | 979 | 71 | ||
| AK166912.1 | - | 9 - 10 | 1013 | 71 | ||
| AK167216.1 | - | 9 - 10 | 1005 | 71 | ||
| AK169532.1 | - | 9 - 10 | 990 | 71 | ||
| BC051639.1 | - | 5 - 6 | 441 | 71 | ||
| BC106089.1 | - | 9 - 10 | 989 | 30715403 | 71 | |
| BC125529.1 | - | 9 - 10 | 978 | 40130044 | 71 | |
| BC125531.1 | - | 9 - 10 | 978 | 40130046 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf4
Genomic Map