Assay Details
| Gene Symbol: Wtip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207212.2 | NP_997095.1 | 4 - 5 | 1014 | 78 | |
| GenBank mRNA | BC042762.1 | - | 4 - 5 | 446 | 78 | |
| BC054125.1 | - | 4 - 5 | 1014 | 6511230 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wtip
Genomic Map