Assay Details
| Gene Symbol: Wtip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207212.2 | NP_997095.1 | 7 - 8 | 1271 | 65 | |
| GenBank mRNA | BC042762.1 | - | 7 - 8 | 703 | 65 | |
| BC054125.1 | - | 7 - 8 | 1271 | 6511230 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
transcription, DNA-templated
regulation of transcription, DNA-templated
cytoskeleton organization
regulation of cell morphogenesis
gene silencing by RNA
gene silencing by miRNA
negative regulation of hippo signaling
negative regulation of nucleic acid-templated transcription
positive regulation of gene silencing by miRNA
transcription corepressor activity
protein binding
zinc ion binding
metal ion binding
response to hypoxia
transcription, DNA-templated
regulation of transcription, DNA-templated
cytoskeleton organization
regulation of cell morphogenesis
gene silencing by RNA
gene silencing by miRNA
negative regulation of hippo signaling
negative regulation of nucleic acid-templated transcription
positive regulation of gene silencing by miRNA
transcription corepressor activity
protein binding
zinc ion binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042342 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042337 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042341 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481686 | BC054125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481686 | NM_207212.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481686 | BC042762.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481686 | BC054125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481686 | NM_207212.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P481686 | BC042762.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40213 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14004 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111178 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155009 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481686 | BC054125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481686 | NM_207212.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481686 | BC042762.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481686 | BC054125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481686 | NM_207212.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P481686 | BC042762.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042342 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155009 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111178 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01494659 | - | - |
| Whole Mouse Genome G4122A | A_52_P481686 | BC054125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481686 | NM_207212.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481686 | BC042762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481686 | BC054125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481686 | NM_207212.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P481686 | BC042762.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100426 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155009 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06631626 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06592210 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06631624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042342 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042337 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06399286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05976102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06605448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05886814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06654238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06618436 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562325 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562330 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433651_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562323 | - | - |
| Mouse Genome 430 2.0 Array | 1433651_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433651_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101543_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101543_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700025209.mm.1 | TC0700002825.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025209.mm.1 | NM_207212.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025209.mm.1 | NM_207212.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025207.mm.1 | TC0700002825.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025207.mm.1 | NM_207212.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025207.mm.1 | NM_207212.2 | Refseq |
| Mouse Expression Set 430 | 1433651_at | - | - |
| Mouse Expression Set 430 | 1433651_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433651_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433651_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 848594 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3830195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2424268 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2424268 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2424268 | - | - |

Wtip
Genomic Map