Assay Details
| Gene Symbol: Pdcd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026549.3 | NP_080825.1 | 4 - 5 | 705 | 95 | |
| GenBank mRNA | AK077948.1 | - | 4 - 5 | 706 | 95 | |
| AK151495.1 | - | 4 - 5 | 709 | 95 | ||
| AK154811.1 | - | 4 - 5 | 721 | 95 | ||
| BC025090.1 | - | 1 - 2 | 169 | 95 | ||
| BC038524.1 | - | 1 - 2 | 175 | 95 | ||
| BC115797.1 | - | 4 - 5 | 704 | 40103840 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdcd2l
Genomic Map