Assay Details
| Gene Symbol: Pdcd2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026549.3 | NP_080825.1 | 6 - 7 | 970 | 95 | |
| GenBank mRNA | AK003339.1 | - | 6 - 7 | 979 | 95 | |
| AK077948.1 | - | 6 - 7 | 971 | 95 | ||
| AK151495.1 | - | 6 - 7 | 974 | 95 | ||
| AK154811.1 | - | 6 - 7 | 986 | 95 | ||
| BC025090.1 | - | 3 - 4 | 434 | 95 | ||
| BC038524.1 | - | 3 - 4 | 440 | 95 | ||
| BC115797.1 | - | 6 - 7 | 969 | 40103840 | 95 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042627 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042629 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042626 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | AK154811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | BC025090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | BC038524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | AK151495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | BC115797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | AK154811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | BC025090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | BC038524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | AK151495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419300 | BC115797.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026380 | AK154811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026380 | BC025090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026380 | BC038524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026380 | AK151495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026380 | BC115797.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623510 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41233 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118593 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129223 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | AK154811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | BC025090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | BC038524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | AK151495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | BC115797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | AK154811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | BC025090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | BC038524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | AK151495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419300 | BC115797.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026380 | AK154811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026380 | BC025090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026380 | BC038524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026380 | AK151495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026380 | BC115797.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25042620 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118593 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129223 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06160076 | - | - |
| Whole Mouse Genome G4122A | A_51_P419300 | AK154811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | BC025090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | BC038524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | AK151495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | BC115797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | AK154811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | BC025090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | BC038524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | AK151495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419300 | BC115797.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118593 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06342268 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06265575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06342273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042626 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042627 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25042629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25042615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06192051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06341890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06275238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06373596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06578425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06342268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06580354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06580353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06086034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05942524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06578423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06275239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25042616 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562354 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562352 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426844_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426845_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562349 | - | - |
| Mouse Genome 430 2.0 Array | 1426845_at | - | - |
| Mouse Genome 430 2.0 Array | 1426845_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426844_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426844_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68079_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68079_at | - | - |
| Mouse Expression Set 430 | 1426844_a_at | - | - |
| Mouse Expression Set 430 | 1426844_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426845_at | - | - |
| Mouse Expression Set 430 | 1426845_PM_at | - | - |
| Murine Genome U74v2 Set | 160448_at | - | - |
| Murine Genome U74v2 Set | 160448_PM_at | - | - |
| Murine Genome U74v2 Set | 168998_at | - | - |
| Murine Genome U74v2 Set | 168998_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426845_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426844_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426844_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426845_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426845_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426845_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426844_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426844_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684300 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684300 | - | - |

Pdcd2l
Genomic Map