Assay Details
| Gene Symbol: Zfp950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029211.1 | - | 5 - 6 | 543 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor kinase 5 | Grk5 | Gprk5 | Anti-sense | Chr. 19 - 60889593 - 61095968 on NCBI Build GRCm38 | |
| Downstream | predicted gene 6020 | Gm6020 |
EG547091, mszf72 |
94.8kb | Anti-sense | Chr. 19 - 61151864 - 61184658 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Zfp950
Genomic Map