Assay Details
| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207270.2 | NP_997153.2 | 9 - 10 | 1906 | 95 | |
| GenBank mRNA | AB217856.1 | - | 8 - 9 | 1579 | 95 | |
| AK078884.1 | - | 8 - 9 | 1583 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprh
Genomic Map