| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10511147 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10511148 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10511146 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400017385.mm.1 |
NR_027971.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017385.mm.1 |
NR_027971.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017385.mm.1 |
NR_027972.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017385.mm.1 |
NR_027972.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017385.mm.1 |
TC0400004206.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400017386.mm.1 |
NR_027971.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017386.mm.1 |
NR_027971.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017386.mm.1 |
TC0400004206.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400017387.mm.1 |
NR_027972.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017387.mm.1 |
NR_027972.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017387.mm.1 |
TC0400004206.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400017384.mm.1 |
NR_027971.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017384.mm.1 |
NR_027971.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017384.mm.1 |
NR_027972.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017384.mm.1 |
NR_027972.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400017384.mm.1 |
TC0400004206.mm.1 |
MTA TC |