Assay Details
| Gene Symbol: Slc35d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177732.4 | NP_808400.1 | 2 - 3 | 250 | 80 | |
| GenBank mRNA | AK049180.1 | - | 2 - 3 | 251 | 80 | |
| AK049462.1 | - | 2 - 3 | 170 | 80 | ||
| AK052968.1 | - | 2 - 3 | 170 | 80 | ||
| AK140305.1 | - | 2 - 3 | 542 | 80 | ||
| AK164517.1 | - | 2 - 3 | 170 | 80 | ||
| AK169775.1 | - | 2 - 3 | 152 | 80 | ||
| AK220330.1 | - | 2 - 3 | 160 | 80 | ||
| BC115984.1 | - | 3 - 4 | 151 | 40105299 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC115984.1 transcripts.
Set Membership: |
|

Slc35d1
Genomic Map