Assay Details
| Gene Symbol: Slc35d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177732.4 | NP_808400.1 | 3 - 4 | 340 | 110 | |
| GenBank mRNA | AK049180.1 | - | 3 - 4 | 341 | 110 | |
| AK049462.1 | - | 3 - 4 | 260 | 110 | ||
| AK052968.1 | - | 3 - 4 | 260 | 110 | ||
| AK140305.1 | - | 3 - 4 | 632 | 110 | ||
| AK164517.1 | - | 3 - 4 | 260 | 110 | ||
| AK169775.1 | - | 3 - 4 | 242 | 110 | ||
| AK220330.1 | - | 3 - 4 | 250 | 110 | ||
| BC115984.1 | - | 4 - 5 | 241 | 40105299 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35d1
Genomic Map