Assay Details
| Gene Symbol: 6030407O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077885.1 | - | 1 - 2 | 351 | 141 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D530049I02 gene | D530049I02Rik | -242.2kb | Anti-sense | Chr. 1 - 73398968 - 73430779 on NCBI Build GRCm38 | |
| Downstream | microRNA 6351 | Mir6351 |
Gm27936, mmu-mir-6351 |
111.5kb | Sense | Chr. 1 - 73855477 - 73855575 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

6030407O03Rik
Genomic Map