Assay Details
| Gene Symbol: 6030407O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045311.1 | 6 - 7 | 1215 | 127 | ||
| GenBank mRNA | AK039327.1 | - | 6 - 7 | 1216 | 127 | |
| AK079660.1 | - | 6 - 7 | 1216 | 127 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6351 | Mir6351 |
Gm27936, mmu-mir-6351 |
Sense | Chr. 1 - 73855477 - 73855575 on NCBI Build GRCm38 | |
| Downstream | tensin 1 | Tns1 |
1110018I21Rik, 1200014E20Rik, AI648117, E030018G17Rik, E030037J05Rik, Tns |
52.1kb | Sense | Chr. 1 - 73910231 - 74124535 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

6030407O03Rik
Genomic Map