Assay Details
| Gene Symbol: 6030407O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045311.1 | 7 - 8 | 1287 | 130 | ||
| GenBank mRNA | AK039327.1 | - | 7 - 8 | 1288 | 130 | |
| AK079660.1 | - | 7 - 8 | 1288 | 130 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D530049I02 gene | D530049I02Rik | -189.4kb | Anti-sense | Chr. 1 - 73398968 - 73430779 on NCBI Build GRCm38 | |
| Downstream | microRNA 6351 | Mir6351 |
Gm27936, mmu-mir-6351 |
182.4kb | Sense | Chr. 1 - 73855477 - 73855575 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

6030407O03Rik
Genomic Map