Assay Details
| Gene Symbol: Hspb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028394.2 | NP_082670.1 | 4 - 5 | 356 | 81 | |
| GenBank mRNA | AK013636.1 | - | 4 - 5 | 333 | 81 | |
| AK157651.1 | - | 4 - 5 | 357 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspb11
Genomic Map