Assay Details
| Gene Symbol: Hspb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028394.2 | NP_082670.1 | 5 - 6 | 393 | 140 | |
| GenBank mRNA | AK013636.1 | - | 5 - 6 | 370 | 140 | |
| AK157651.1 | - | 5 - 6 | 394 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23189558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23189557 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024744 | AK135163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024744 | BC021897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024744 | BC034465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024749 | AK157651.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024749 | NM_028394.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024749 | AK013636.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5595 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483689 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159478 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116954 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161448 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024749 | AK157651.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024749 | NM_028394.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024749 | AK013636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024744 | AK135163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024744 | BC021897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024744 | BC034465.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00954417 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161448 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116954 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00954424 | - | - |
| Whole Mouse Genome G4122A | A_51_P345685 | AK157651.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345685 | NM_028394.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345685 | AK013636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485918 | AK135163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485918 | BC021897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485918 | BC034465.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161448 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116954 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03948130 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03948138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23189557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23189558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23189583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03683895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03411831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03568785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03275616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03948128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03935984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03568784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03948139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03338296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03301331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03979363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03683891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03825322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03575891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23189674 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10506661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506660 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506667 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506666 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506663 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453016_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425935_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10506658 | - | - |
| Mouse Genome 430 2.0 Array | 1425935_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453016_at | - | - |
| Mouse Genome 430 2.0 Array | 1453016_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425935_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72938_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72938_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400007794.mm.1 | NM_028394.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400007794.mm.1 | TC0400001125.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400007793.mm.1 | NM_028394.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400007793.mm.1 | TC0400001125.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425935_at | - | - |
| Mouse Expression Set 430 | 1453016_at | - | - |
| Mouse Expression Set 430 | 1425935_PM_at | - | - |
| Mouse Expression Set 430 | 1453016_PM_at | - | - |
| Murine Genome U74v2 Set | 104085_PM_at | - | - |
| Murine Genome U74v2 Set | 104085_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425935_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453016_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453016_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425935_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425935_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453016_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425935_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453016_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 430195 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2744108 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2744108 | - | - |

Hspb11
Genomic Map