Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 7 - 8 | 895 | 88 | |
| NM_198438.1 | NP_940840.1 | 6 - 7 | 814 | 88 | ||
| GenBank mRNA | AF170906.1 | - | 6 - 7 | 544 | 88 | |
| AF500117.1 | - | 6 - 7 | 544 | 88 | ||
| AK011509.1 | - | 7 - 8 | 513 | 88 | ||
| AK011853.1 | - | 7 - 8 | 865 | 88 | ||
| AK017711.1 | - | 7 - 8 | 898 | 88 | ||
| AK133812.1 | - | 4 - 5 | 348 | 89 | ||
| AK153733.1 | - | 3 - 4 | 255 | 88 | ||
| AK211107.1 | - | 4 - 5 | 264 | 88 | ||
| AY167987.1 | - | 6 - 7 | 427 | 88 | ||
| BC003430.1 | - | 6 - 7 | 568 | 2646149 | 88 | |
More Information
Set Membership: |
|

Ssbp3
Genomic Map