Assay Details
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252654.1 | NP_001239583.1 | 13 - 14 | 1094 | 87 | |
| NM_001252655.1 | NP_001239584.1 | 13 - 14 | 1123 | 87 | ||
| NM_001252656.1 | NP_001239585.1 | 14 - 15 | 1165 | 87 | ||
| NM_010598.3 | NP_034728.2 | 14 - 15 | 1136 | 87 | ||
| GenBank mRNA | AK143166.1 | - | 13 - 14 | 1071 | 87 | |
| AK157053.1 | - | 13 - 14 | 1087 | 87 | ||
| BC039178.1 | - | 14 - 15 | 1066 | 4503373 | 87 | |
| L48983.1 | - | 14 - 15 | 1128 | 87 | ||
| U31908.1 | - | 14 - 15 | 1073 | 87 | ||
| U65592.1 | - | 13 - 14 | 931 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab2
Genomic Map