Assay Details
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252656.1 | NP_001239585.1 | 2 - 3 | 304 | 86 | |
| NM_010598.3 | NP_034728.2 | 2 - 3 | 275 | 86 | ||
| GenBank mRNA | AK152613.1 | - | 2 - 3 | 227 | 86 | |
| BC039178.1 | - | 2 - 3 | 205 | 4503373 | 86 | |
| L48983.1 | - | 2 - 3 | 267 | 86 | ||
| U31908.1 | - | 2 - 3 | 212 | 86 | ||
| U65592.1 | - | 1 - 2 | 70 | 86 | ||
More Information
Set Membership: |
|

Kcnab2
Genomic Map