Assay Details
| Gene Symbol: Acot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146057.1 | NP_001139529.1 | 7 - 8 | 940 | 60 | |
| NM_001146058.1 | NP_001139530.1 | 7 - 8 | 802 | 60 | ||
| NM_133348.2 | NP_579926.2 | 7 - 8 | 929 | 60 | ||
| GenBank mRNA | AB049821.1 | - | 7 - 8 | 895 | 60 | |
| AB088411.1 | - | 7 - 8 | 868 | 60 | ||
| AB088412.1 | - | 7 - 8 | 836 | 60 | ||
| AB207243.1 | - | 7 - 8 | 895 | 60 | ||
| AK010646.1 | - | 759 | 60 | |||
| AK152277.1 | - | 8 - 9 | 810 | 60 | ||
| BC013507.1 | - | 7 - 8 | 797 | 4224774 | 60 | |
More Information
Set Membership: |
|

Acot7
Genomic Map