Assay Details
| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076209.1 | - | 1 - 2 | 74 | 89 | |
| AK136208.1 | - | 1 - 2 | 70 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076209.1, AK136208.1 transcripts.
Set Membership: |
|

Oma1
Genomic Map