Assay Details
| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025909.3 | NP_080185.1 | 6 - 7 | 1182 | 75 | |
| GenBank mRNA | AK008020.1 | - | 6 - 7 | 1183 | 75 | |
| AK076209.1 | - | 6 - 7 | 1212 | 75 | ||
| BC016238.1 | - | 5 - 6 | 1137 | 4504835 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oma1
Genomic Map