Assay Details
| Gene Symbol: Nr3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155200.1 | - | 1 - 2 | 104 | 143 | |
| BC016462.1 | - | 1 - 2 | 105 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155200.1, BC016462.1 transcripts.
Set Membership: |
|

Nr3c1
Genomic Map