Assay Details
| Gene Symbol: Nr3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008173.3 | NP_032199.3 | 6 - 7 | 2072 | 119 | |
| GenBank mRNA | AK037733.1 | - | 6 - 7 | 1953 | 119 | |
| AK155200.1 | - | 8 - 9 | 2154 | 119 | ||
| AK156323.1 | - | 8 - 9 | 2148 | 119 | ||
| BC129912.1 | - | 7 - 8 | 2050 | 40111802 | 119 | |
| BC129913.1 | - | 7 - 8 | 2050 | 40111803 | 119 | |
| DQ504162.1 | - | 6 - 7 | 2056 | 119 | ||
| HM236293.1 | - | 7 - 8 | 2032 | 119 | ||
| X04435.1 | - | 7 - 8 | 2050 | 119 | ||
| X13358.1 | - | 7 - 8 | 2032 | 119 | ||
| X13359.1 | - | 7 - 8 | 2035 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr3c1
Genomic Map