Assay Details
| Gene Symbol: Dhcr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053272.2 | NP_444502.2 | 3 - 4 | 625 | 58 | |
| GenBank mRNA | AK017937.1 | - | 3 - 4 | 387 | 58 | |
| AK083737.1 | - | 3 - 4 | 580 | 58 | ||
| AY039762.1 | - | 3 - 4 | 513 | 58 | ||
| BC019797.1 | - | 3 - 4 | 569 | 5054108 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhcr24
Genomic Map