Assay Details
| Gene Symbol: Dhcr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053272.2 | NP_444502.2 | 6 - 7 | 1153 | 77 | |
| GenBank mRNA | AK017937.1 | - | 6 - 7 | 915 | 77 | |
| AK083737.1 | - | 6 - 7 | 1108 | 77 | ||
| AK129036.1 | - | 6 - 7 | 1153 | 77 | ||
| AY039762.1 | - | 6 - 7 | 1047 | 77 | ||
| BC019797.1 | - | 6 - 7 | 1097 | 5054108 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhcr24
Genomic Map