Assay Details
| Gene Symbol: Tle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285529.1 | NP_001272458.1 | 10 - 11 | 1647 | 126 | |
| NM_001285530.1 | NP_001272459.1 | 8 - 9 | 1428 | 126 | ||
| NM_011599.5 | NP_035729.3 | 10 - 11 | 1650 | 126 | ||
| GenBank mRNA | AK048515.1 | - | 10 - 11 | 1637 | 126 | |
| AK052961.1 | - | 8 - 9 | 1751 | 126 | ||
| AK082499.1 | - | 10 - 11 | 1616 | 126 | ||
| AY155197.1 | - | 8 - 9 | 586 | 126 | ||
| AY155198.1 | - | 6 - 7 | 449 | 126 | ||
| AY155199.1 | - | 10 - 11 | 126 | |||
| AY155200.1 | - | 4 - 5 | 283 | 126 | ||
| BC057573.1 | - | 10 - 11 | 1477 | 6401650 | 126 | |
| U61362.1 | - | 10 - 11 | 816 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tle1
Genomic Map