Assay Details
| Gene Symbol: Tro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002272.3 | NP_001002272.1 | 3 - 4 | 1791 | 98 | |
| NM_001290770.1 | NP_001277699.1 | 3 - 4 | 1740 | 98 | ||
| NM_001290771.1 | NP_001277700.1 | 3 - 4 | 1791 | 98 | ||
| NM_001290772.1 | NP_001277701.1 | 3 - 4 | 1791 | 98 | ||
| NM_019548.4 | NP_062421.2 | 3 - 4 | 1791 | 98 | ||
| GenBank mRNA | AB017108.1 | - | 1 - 2 | 132 | 98 | |
| AB032477.1 | - | 3 - 4 | 1659 | 98 | ||
| AB036783.1 | - | 4 - 5 | 1733 | 98 | ||
| AF241244.1 | - | 3 - 4 | 1655 | 98 | ||
| AF241245.1 | - | 3 - 4 | 1655 | 98 | ||
| AF288605.1 | - | 4 - 5 | 1733 | 98 | ||
| AF288606.1 | - | 4 - 5 | 1733 | 98 | ||
| AF319977.1 | - | 2 - 3 | 1674 | 98 | ||
| AK122449.1 | - | 2 - 3 | 681 | 98 | ||
| BC075630.1 | - | 3 - 4 | 1740 | 30536880 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF241244.1 transcripts.
Set Membership: |
|

Tro
Genomic Map