Assay Details
| Gene Symbol: Pepd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008820.2 | NP_032846.2 | 4 - 5 | 428 | 88 | |
| GenBank mRNA | AK010581.1 | - | 4 - 5 | 409 | 88 | |
| AK145843.1 | - | 4 - 5 | 459 | 88 | ||
| AK168384.1 | - | 4 - 5 | 418 | 88 | ||
| BC086644.1 | - | 4 - 5 | 402 | 6807359 | 88 | |
| BC131945.1 | - | 4 - 5 | 430 | 40130222 | 88 | |
| BC131973.1 | - | 4 - 5 | 430 | 40130250 | 88 | |
| D82983.1 | - | 4 - 5 | 415 | 88 | ||
| U51014.1 | - | 4 - 5 | 451 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pepd
Genomic Map