Assay Details
| Gene Symbol: Oacyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177028.3 | NP_796002.1 | 2 - 3 | 319 | 60 | |
| GenBank mRNA | AK030604.1 | - | 2 - 3 | 319 | 60 | |
| BC141265.1 | - | 2 - 3 | 316 | 9055877 | 60 | |
More Information
Set Membership: |
|

Oacyl
Genomic Map