Assay Details
| Gene Symbol: Fhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289533.1 | NP_001276462.1 | 1 - 2 | 105 | 85 | |
| GenBank mRNA | AK052936.1 | - | 1 - 2 | 106 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052936.1, NM_001289533.1 transcripts.
Set Membership: |
|

Fhl2
Genomic Map