Assay Details
| Gene Symbol: Pfkfb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008824.3 | NP_032850.1 | 1 - 2 | 163 | 136 | |
| GenBank mRNA | AK132629.1 | - | 1 - 2 | 164 | 136 | |
| BC132561.1 | - | 1 - 2 | 148 | 40130838 | 136 | |
| BC132563.1 | - | 1 - 2 | 148 | 40130840 | 136 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132629.1, BC132561.1, BC132563.1, NM_008824.3 transcripts.
Set Membership: |
|

Pfkfb1
Genomic Map