Assay Details
| Gene Symbol: Bcl2l14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025778.3 | NP_080054.1 | 2 - 3 | 246 | 69 | |
| GenBank mRNA | AK016670.1 | - | 2 - 3 | 247 | 69 | |
| AK016997.1 | - | 2 - 3 | 246 | 69 | ||
| AK160247.1 | - | 2 - 3 | 249 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016670.1, AK016997.1, AK160247.1, NM_025778.3 transcripts.
Set Membership: |
|

Bcl2l14
Genomic Map