Assay Details
| Gene Symbol: Ccdc186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170757.1 | NP_739563.1 | 2 - 3 | 815 | 104 | |
| GenBank mRNA | AF349751.1 | - | 2 - 3 | 815 | 104 | |
| AK041398.1 | - | 3 - 4 | 1002 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc186
Genomic Map