Assay Details
| Gene Symbol: Ccdc186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170757.1 | NP_739563.1 | 4 - 5 | 1153 | 113 | |
| GenBank mRNA | AF349751.1 | - | 4 - 5 | 1153 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc186
Genomic Map