Assay Details
| Gene Symbol: Acsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027976.2 | NP_082252.1 | 5 - 6 | 1261 | 129 | |
| GenBank mRNA | AK034942.1 | - | 1261 | 129 | ||
| AK145894.1 | - | 5 - 6 | 922 | 129 | ||
| AK150542.1 | - | 5 - 6 | 927 | 129 | ||
| AK150663.1 | - | 5 - 6 | 927 | 129 | ||
| AK151378.1 | - | 5 - 6 | 716 | 129 | ||
| AK161024.1 | - | 5 - 6 | 614 | 129 | ||
| BC031544.1 | - | 5 - 6 | 695 | 3987201 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl5
Genomic Map