Assay Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008149.3 | NP_032175.2 | 10 - 11 | 1263 | 66 | |
| GenBank mRNA | AK047295.1 | - | 10 - 11 | 1264 | 66 | |
| AK137067.1 | - | 12 - 13 | 1544 | 66 | ||
| AK173202.1 | - | 10 - 11 | 1288 | 66 | ||
| BC019201.1 | - | 10 - 11 | 1268 | 5041120 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpam
Genomic Map