Assay Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008149.3 | NP_032175.2 | 1 - 2 | 125 | 96 | |
| GenBank mRNA | BC019201.1 | - | 1 - 2 | 130 | 5041120 | 96 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC019201.1, NM_008149.3 transcripts.
Set Membership: |
|

Gpam
Genomic Map