Assay Details
| Gene Symbol: Vwa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172840.2 | NP_766428.2 | 11 - 12 | 1739 | 65 | |
| GenBank mRNA | AJ536329.1 | - | 11 - 12 | 1739 | 65 | |
| AK029297.1 | - | 11 - 12 | 1686 | 65 | ||
| AK133486.1 | - | 11 - 12 | 1646 | 65 | ||
| AK145058.1 | - | 11 - 12 | 1909 | 65 | ||
| AK160971.1 | - | 11 - 12 | 1745 | 65 | ||
| AK166339.1 | - | 11 - 12 | 1818 | 65 | ||
| BC115868.1 | - | 10 - 11 | 1641 | 40081354 | 65 | |
| BC116636.1 | - | 11 - 12 | 1689 | 40081359 | 65 | |
More Information
Set Membership: |
|

Vwa2
Genomic Map